Chloroform-[D₁] Isotope_(99.8% D) NMR

Supplier: VWR Chemicals
Warning

Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

87153.0100EA 112 GBP
87153.0100 87153.0006 87153.0500
Chloroform-[D₁] Isotope_(99.8% D) NMR
Chloroform-[D1]
Formula: CDCl₃
MW: 120.38 g/mol
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64,1 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1 III

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.80 %
Water Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR