VWR® AVS TITRINORM, Cerium Standard Solution, 1000 mg/L Ce in dil. Nitric Acid, Standard for AAS
Supplier: VWR Chemicals
86669.180EA 42.8 GBP
86669.180 86669.260
VWR® AVS TITRINORM, Cerium Standard Solution, 1000 mg/L Ce in dil. Nitric Acid, Standard for AAS
Standards Cationic Standards Cerium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%
Order Now
SPECIFICATIONS
Environmentally Preferable