Silver sulphate 80 g/l sulphuric acid_96, AVS TITRINORM COD_determination_DIN 38409-H 43-1

Supplier: VWR Chemicals
Danger

Synonyms: Disilver sulphate

86189.290EA 340 GBP
86189.290
Silver sulphate 80 g/l sulphuric acid_96, AVS TITRINORM COD_determination_DIN 38409-H 43-1
Silver sulphate
Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085
Melting Pt: 655
MDL Number: MFCD00003407
CAS Number: 10294-26-5
UN: 3264
ADR: 8 II
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Composition complies with DIN 38409 part 43 - DEV H43

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR