n-Pentane 99,5 GC anhydrous basis, PESTINORM® Supra Trace

Supplier: VWR Chemicals
Danger

Synonyms: Pentane

Filter 0,2 um_Inert_Gas_filled

85852.290EA 110 GBP
85852.290 85852.320
n-Pentane 99,5 GC anhydrous basis, PESTINORM® Supra Trace
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless liquid
Assay (GC, on anhydrous basis) min. 99.5 %
Acidity (as Acetic acid) max. 0.002 %
Color (APHA, Pt-Co) max. 5
Residue after evaporation max. 0.0001 % w/w
Water (KF) max. 0.005 % w/w
GC/ECD Dioxins, Furans & PCB's max. 5 pg/ml
GC/ECD any Pesticide (as Lindane) max. 2 pg/ml
GC/NPD any Pesticide (as Parathion) max. 5 pg/ml
GC/ECD 1,2,4-TCB to daca-PCB (as Lindane) max. 5 pg/ml
GC/ECD DCM to 1,2,4-TCB (as TCM) max. 2 ng/ml
GC/FID C10 to C40 (as n-Decane) max. 2 ng/ml
GC/MSD C10 to C40 (as Decane; 30-600 amu) max. 2 ng/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR