Methanol 99,9, HiPerSolv CHROMANORM®, ultra LC-MS, UPLC/UHPLC-MS suitable

Supplier: VWR Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filter 0,2 um_Nitrogen_filled

85800.290EA 30.2 GBP
85800.290 85800.320
Methanol 99,9, HiPerSolv CHROMANORM®, ultra LC-MS, UPLC/UHPLC-MS suitable
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.9 %
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0002 meq/g
Boiling point 64 - 65 °C
Colouration Max. 10 APHA
Density (20/20) 0.791 - 0.793
Evaporation residue Max. 1 ppm
Water Max. 0.02 %
Al (Aluminium) Max. 20 ppb
Ag (Silver) Max. 100 ppb
Ba (Barium) Max. 100 ppb
Ca (Calcium) Max. 200 ppb
Cd (Cadmium) Max. 20 ppb
Co (Cobalt) Max. 20 ppb
Cu (Copper) Max. 10 ppb
Fe (Iron) Max. 50 ppb
K (Potassium) Max. 50 ppb
Mg (Magnesium) Max. 50 ppb
Mn (Manganese) Max. 100 ppb
Mo (Molybdenum) Max. 20 ppb
Na (Sodium) Max. 200 ppb
Ni (Nickel) Max. 100 ppb
Pb (Lead) Max. 20 ppb
Zn (Zinc) Max. 100 ppb
Gradient (220 nm) Max. 3 mAU
Gradient (235 nm) Max. 2 mAU
Gradient (254 nm) Max. 1 mAU
Transmittance (210 nm) Min. 45 %
Transmittance (220 nm) Min. 65 %
Transmittance (230 nm) Min. 85 %
Transmittance (235 nm) Min. 90 %
Transmittance (240 nm) Min. 95 %
Transmittance (250 nm) Min. 95 %
Transmittance (260 nm) Min. 98 %
Transmittance (280-400 nm) Min. 98 %
Absorbance (210 nm) Max. 0.347
Absorbance (220 nm) Max. 0.188
Absorbance (225 nm) Max. 0.155
Absorbance (230 nm) Max. 0.071
Absorbance (240 nm) Max. 0.023
Absorbance (250 nm) Max. 0.023
Absorbance (260 nm) Max. 0.009
Absorbance (280-400 nm) Max. 0.009
Fluorescence (as quinine; 254 nm) Max. 1 ppb
Fluorescence (as quinine) (365 nm) Max. 0.5 ppb
Conforms Ph.Eur. R1 1053201 Passes test
Conforms Ph.Eur. R2 1053202 Passes test
Conforms to ACS Passes test
Filtered at 0.2 µm Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR