VWR®, Palladium standard solutions for ICP-MS, ARISTAR®
Supplier: VWR Chemicals
85587.180EA 243 GBP
85587.180 457371K
VWR®, Palladium standard solutions for ICP-MS, ARISTAR®
Standards Cationic Standards Palladium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0.5% (typical)
  • Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities
Order Now
SPECIFICATIONS
Environmentally Preferable