Acetonitrile, Wasser_0,001 99,95 DNA_synthesis

Supplier: VWR Chemicals
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

Filter 0,2 um_Nitrogen_filled

85501.290EA 110 GBP
85501.290 85501.400 85501.320 85501.500 85501.911 85501.914 VWRC85501.321
Acetonitrile, Wasser_0,001 99,95 DNA_synthesis
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.95 %
IR Spectrum Passes test
Acidity Max. 0.001 meq/g
Alkalinity Max. 0.0002 meq/g
Colouration Max. 10 APHA
Evaporation residue Max. 1 ppm
Water Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR