Sodium hydroxide 6.25 mol/l (6.25 N) aqueous solution

Supplier: VWR Collection
Danger

Synonyms: Caustic soda

85100.360EA 0 GBP
85100.360
Sodium hydroxide 6.25 mol/l (6.25 N) aqueous solution
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR