Acetic acid 99% 99, NORMATOM®, ultrapure Trace_metal

Supplier: VWR Chemicals
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

85030.270EA 830 GBP
85030.270 85030.230
Acetic acid 99% 99, NORMATOM®, ultrapure Trace_metal
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3) II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99%
Ag (Silver) 50 ppt
Al (Aluminium) 50 ppt
As (Arsenic) 50 ppt
Ba (Barium) 10 ppt
Be (Beryllium) 10 ppt
Bi (Bismuth) 10 ppt
Ca (Calcium) 50 ppt
Cd (Cadmium) 10 ppt
Ce (Cerium) 10 ppt
Co (Cobalt) 10 ppt
Cr (Chromium) 10 ppt
Cs (Cesium) 10 ppt
Cu (Copper) 10 ppt
Dy (Dysprosium) 1 ppt
Er (Erbium) 1 ppt
Eu (Europium) 1 ppt
Fe (Iron) 50 ppt
Ga (Gallium) 10 ppt
Gd (Gadolinium) 1 ppt
Ge (Germanium) 10 ppt
Hf (Hafnium) 10 ppt
Ho (Holmium) 1 ppt
In (Indium) 1 ppt
K (Potassium) 50 ppt
La (Lanthanum) 1 ppt
Li (Lithium) 10 ppt
Lu (Lutetium) 10 ppt
Mg (Magnesium) 50 ppt
Mn (Manganese) 10 ppt
Mo (Molybdenum) 10 ppt
Na (Sodium) 100 ppt
Nd (Neodymium) 1 ppt
Ni (Nickel) 50 ppt
Pb (Lead) 10 ppt
Pr (Praseodymium) 1 ppt
Pt (Platinum) 50 ppt
Rb (Rubidium) 10 ppt
Re (Rhenium) 10 ppt
Rh (Rhodium) 50 ppt
Ru (Ruthenium) 50 ppt
Sb (Antimony) 50 ppt
Sc (Scandium) 10 ppt
Se (Selenium) Information only
Sm (Samarium) 1 ppt
Sn (Tin) 50 ppt
Sr (Strontium) 10 ppt
Tb (Terbium) 1 ppt
Te (Tellurium) 1 ppt
Th (Thorium) 1 ppt
Ti (Titanium) 10 ppt
Tl (Thallium) 10 ppt
Tm (Thulium) 1 ppt
U (Uranium) 1 ppt
V (Vanadium) 10 ppt
W (Tungsten) 10 ppt
Y (Yttrium) 1 ppt
Yb (Ytterbium) 1 ppt
Zn (Zinc) 50 ppt
Zr (Zirconium) 10 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR