N-Methyl-2-pyrrolidone (NMP) 99,5 Peptide_synthesis

Supplier: VWR Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

84572.320EA 162 GBP
84572.320
N-Methyl-2-pyrrolidone (NMP) 99,5 Peptide_synthesis
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
REACH: 01-2119472430-46
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 99.5 %
Appearance Clear colourless liquid
Colour value Max. 10 APHA
Free amines Max. 0.001 %
Residue on evaporation Max. 0.002 %
Water Max. 0.02 %
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Pb (Lead) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR