Toluene 99,7, PESTINORM® Pest_Residual

Supplier: VWR Chemicals
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

Filter 0,2 um_Nitrogen_filled

83664.320EA 141 GBP
83664.320 83664.400
Toluene 99,7, PESTINORM® Pest_Residual
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 01-2119471310-51
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.7 %
Evaporation residue Max. 5 ppm
Water Max. 0.03 %
Lindane (GC/ECD) ≤ 5 ng/l Passes test
Pesticide analysis (Lindane/ECD) Max. 5 ng/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR