2,2,4-Trimethylpentane 99,5, HiPerSolv CHROMANORM® HPLC

Supplier: VWR Chemicals
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

Filter 0,2 um_Nitrogen_filled

83630.320EA 302 GBP
83630.320 83630.290
2,2,4-Trimethylpentane 99,5, HiPerSolv CHROMANORM® HPLC
2,2,4-Trimethylpentane
Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.50 %
Water Max. 0.01 %
Non-volatile residue Max. 0.0005 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Transmittance (220 nm) Min. 70.0 %
Transmittance (235 nm) Min. 80.0 %
Transmittance (255 nm) Min. 98.0 %
Conforms to BDH 15246 Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR