Chloroform 99,8 Stabilised, HiPerSolv CHROMANORM® HPLC

Supplier: VWR Chemicals
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilisation: 20 ppm 2-Methyl-2-buten
Filter 0,2 um_Nitrogen_filled

83626.320EA 204 GBP
83626.320 83626.290
Chloroform 99,8 Stabilised, HiPerSolv CHROMANORM® HPLC
Chloroform
Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 01-2119486657-20
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.80 %
Water Max. 0.0500 %
Non-volatile residue Max. 0.001 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Transmittance (250 nm) Min. 50.0 %
Transmittance (260 nm) Min. 84.00 %
Transmittance (280 nm) Min. 95.0 %
Transmittance (300 nm) Min. 98.0 %
Conforms to BDH 15283 Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR