Pyridine ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Azabenzene, Azine

8226.2500EA 384 GBP
8226.2500
Pyridine ≥99%, GPR RECTAPUR®
Pyridine
Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Evaporation residue Max. 20 ppm
Water Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR