Labelling tapes, set
Supplier: PRECISION DYNAMICS TIMEM
T-501-RP T-512-RP T-534-RP
817-0026EA 249 GBP
817-0026 817-0027 817-0044
Labelling tapes, set
Tapes
This all-purpose, self-adhesive tape is well suited for a wide variety of labelling applications. It can be written on with pen, pencil or permanent marker and is available in various colours, in three widths.
  • Temperature resistant from −45 to +121 °C
  • With removable adhesive
  • Oil, water and acid resistant
Set 1 contains two rolls each of white, yellow, blue, green, and red, and one roll of orange and pink.
Set 2 contains five rolls of white, four rolls of yellow, and three rolls each of blue, green, orange, red and pink.
Set 3 contains four rollsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable