Polyethylene glycol 1,500, Supelco®

Supplier: Merck
Warning

Synonyms: PEG 1500, PEG 1,500

MQ100

81289-1G
81289-1GEA 190 GBP
81289-1G
Polyethylene glycol 1,500, Supelco®
Polyethylene glycol 1,500
Formula: H(OCH₂CH₂)nOH
MW: 1500 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 44…48 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 260
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b2930ddb0dad09b0d006cd81d2ef8b4f00f2e87&vid=324418b150d3b280a75d92ae7726249505176dc9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR