Acetone, BAKER, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

8003.1000EA 36.5 GBP
8003.1000 8003.2500 8003.5000 8003.9025 8003.9010 8003.9200ST
Acetone, BAKER, J.T.Baker®
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Acidity or Alkalinity passes test
Appearance of solution passes test
Identification passes test
Matter Insoluble in Water passes test
Reducing Substances passes test
Relative Density 0.790 - 0.793
Residue on Evaporation max. 50 ppm
Water (H2O) max. 3 g/l
Related Substances:
Benzene max. 2 ppm (v/v)
Methanol (CH3OH) max. 0.05 % (v/v)
Other Impurities passes test
2-Propanol (CH3CHOHCH3) max. 0.05 % (v/v)
Additional Specification(s):
Assay (by GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR