N-Ethyl-o-toluidine zS, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-(Ethylamino)toluene, N-Ethyl-2-methylaniline

MQ200

8.43602.0100 8.43602.0500
8.43602.0100EA 65.9 GBP
8.43602.0100 8.43602.0500
N-Ethyl-o-toluidine zS, Sigma-Aldrich®
N-Ethyl-o-toluidine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 212…212.5 °C (1013 hPa)
Melting Pt: –15 °C
Density: 0.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00026695
CAS Number: 94-68-8
EINECS: 202-354-3
UN: 2754
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR