3-(Dimethylamino)propiophenone hydrochloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

8.41828.0025
8.41828.0025EA 72.4 GBP
8.41828.0025
3-(Dimethylamino)propiophenone hydrochloride, Sigma-Aldrich®
3-(Dimethylamino)propiophenone hydrochloride
Formula: C₁₁H₁₆ClNO
MW: 213.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012481
CAS Number: 879-72-1
EINECS: 212-909-1
UN: 2811

Order Now

Specification Test Results

Melting Point 155 - 156 °C
Molar Mass 213.7 g/mol
Hill Formula C11H16ClNO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR