Tetrapentylammonium iodide zS, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Tetra-n-pentylammonium iodide, Tetraamylammonium iodide

MQ200

8.41762.0025
8.41762.0025EA 155 GBP
8.41762.0025
Tetrapentylammonium iodide zS, Sigma-Aldrich®
Tetrapentylammonium iodide
Formula: C₂₀H₄₄IN
MW: 425.48 g/mol
Melting Pt: 135…137 °C
Density: 1.18 g/cm³ (25 °C)
MDL Number: MFCD00041980
CAS Number: 2498-20-6
EINECS: 219-688-0

Order Now

Specification Test Results

Solubility 7.40 g/l (25 °C)
Melting Point 135 - 137 °C
Molar Mass 425.47 g/mol

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR