(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) zS, Sigma-Aldrich®

Supplier: Merck

Synonyms: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

MQ200

8.41747.0001
8.41747.0001EA 48.3 GBP
8.41747.0001
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) zS, Sigma-Aldrich®
(2-Nitrophenyl) β-D-galactopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063255
CAS Number: 369-07-3
EINECS: 206-716-1

Order Now

Specification Test Results

Molar Mass 301.25 g/mol
Hill Formula C12H15NO8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR