L(-)-Carnitine 98,0 zS pH 6.5 - 8.5 (50 g/l, H₂O), Sigma-Aldrich®

Supplier: Merck

Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate

MQ200

8.40092.0025
8.40092.0025EA 127 GBP
8.40092.0025
L(-)-Carnitine 98,0 zS pH 6.5 - 8.5 (50 g/l, H₂O), Sigma-Aldrich®
L(-)-Carnitine
Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
MDL Number: MFCD00038747
CAS Number: 541-15-1
EINECS: 208-768-0

Order Now

Specification Test Results

Solubility 800 g/l (20 °C)
Melting Point 190 - 195 °C decomposes
Molar Mass 161.2 g/mol
Bulk Density 636 kg/m3
Density 0.64 g/cm3
pH 6.5 - 8.5 (50 g/l, H2O)
Ignition Point 365 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR