1-Butanol, EMPLURA®, Supelco®

Supplier: Merck
Danger

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

MQ200

8.22262.2500
8.22262.2500EA 68.85 GBP
8.22262.2500
1-Butanol, EMPLURA®, Supelco®
1-Butanol
Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6
Melting Pt: –89 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 30
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Limited Time Sale

Select items below are currently on sale

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR