n-Docosane zS, Sigma-Aldrich®

Supplier: Merck

Synonyms: N-docosane

MQ200

8.22046.0050
8.22046.0050EA 59.2 GBP
8.22046.0050
n-Docosane zS, Sigma-Aldrich®
n-Docosane
Formula: H₃C(CH₂)₂₀CH₃
MW: 310.61 g/mol
Boiling Pt: 368
Melting Pt: 44
Density: 0.7944 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009348
CAS Number: 629-97-0
EINECS: 211-121-5

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 44 °C
Molar Mass 310.6 g/mol
Boiling Point 369 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 110 °C
Density 0.794 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR