1-Octene zS, Sigma-Aldrich®

Supplier: Merck
Danger


MQ200

8.20928.0005 8.20928.0250 8.20928.1000
8.20928.0005EA 28.3 GBP
8.20928.0005 8.20928.0250 8.20928.1000
1-Octene zS, Sigma-Aldrich®
1-Octene
Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 121
Melting Pt: –102 °C
Density: 0.7152 g/cm³ (20 °C)
Flash Pt: 8
Storage Temperature: Ambient
MDL Number: MFCD00009548
CAS Number: 111-66-0
EINECS: 203-893-7
UN: 3295
ADR: 3 II
Merck Index: 13,01770

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point -102 °C
Molar Mass 112.21 g/mol
Boiling Point 121 °C
Vapor Pressure 17.3 hPa (20 °C)
Flash Point 8 °C
Refractive Index 1.4085 (20 °C, 589 nm)
Explosion Limit 0.7 - 6.8 %(V)
Density 0.72 g/cm3 (20 °C)
Ignition Point 221 °C
Viscosity kinematic 0.38 mm2/s (40 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR