2-Methylhydroquinone zS, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Methylhydroquinone

MQ200

8.20801.0100
8.20801.0100EA 55 GBP
8.20801.0100
2-Methylhydroquinone zS, Sigma-Aldrich®
2-Methylhydroquinone
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 285
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002345
CAS Number: 95-71-6
EINECS: 202-443-7
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Solubility 77 g/l (25 °C)
Melting Point 124 - 127 °C
Molar Mass 124.14 g/mol
Boiling Point 285 °C
Flash Point 172 °C
Ignition Point 468 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR