Ethylenediammonium sulphate ≥97% for synthesis, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1.2-Diaminoethane sulphate

M-Clarity™ quality level = MQ200

8.20031.0250
8.20031.0250EA 119 GBP
8.20031.0250
Ethylenediammonium sulphate ≥97% for synthesis, Sigma-Aldrich®
Ethylenediammonium sulphate
Formula: C₂H₈N₂
MW: 158.17 g/mol
MDL Number: MFCD00036417
CAS Number: 22029-36-3
EINECS: 206-369-6

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 225 - 228 °C
Molar Mass 158.17 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR