4,4-Dimethyl-2-oxazoline zS, Sigma-Aldrich®

Supplier: Merck
Danger


MQ200

8.18107.0005
8.18107.0005EA 150 GBP
8.18107.0005
4,4-Dimethyl-2-oxazoline zS, Sigma-Aldrich®
4,4-Dimethyl-2-oxazoline
Formula: C₅H₉NO
MW: 99.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014507
CAS Number: 30093-99-3
EINECS: 250-045-7
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 99.13 g/mol
Boiling Point 99 - 100 °C (1013 hPa)
Flash Point 1 °C
Density 0.94 g/cm3 (20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR