Dodecyltrimethylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: (1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC

M-Clarity™ quality level = MQ200

8.16142.0025 8.16142.1000
8.16142.0025EA 124 GBP
8.16142.0025 8.16142.1000
Dodecyltrimethylammonium chloride for synthesis, Sigma-Aldrich®
Dodecyltrimethylammonium chloride
Formula: C₁₅H₃₄ClN
MW: 263.89 g/mol
Melting Pt: 248…249.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00041974
CAS Number: 112-00-5
EINECS: 203-927-0
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Melting Point 237 °C (decomposition)
Molar Mass 263.9 g/mol
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR