Sulphuryl chloride for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Sulphuryl dichloride

M-Clarity™ quality level = MQ200

8.07997.0250 8.07997.1000 8.07997.9300
8.07997.0250EA 90.8 GBP
8.07997.0250 8.07997.1000 8.07997.9300
Sulphuryl chloride for synthesis, Sigma-Aldrich®
Sulphuryl chloride
Formula: Cl₂O₂S
MW: 134.97 g/mol
Boiling Pt: 69.1
Melting Pt: –54 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011451
CAS Number: 7791-25-5
EINECS: 232-245-6
UN: 1834
ADR: 6.1 (8) I
Merck Index: 12,09153

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR