Iron(III) chloride, anhydrous zS, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Iron trichloride, Ferric chloride

MQ200

8.03945.0500 8.03945.1000 8.03945.9060 8.03945.9025
8.03945.0500EA 53 GBP
8.03945.0500 8.03945.1000 8.03945.9060 8.03945.9025
Iron(III) chloride, anhydrous zS, Sigma-Aldrich®
Iron (III) chloride
Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 315
Melting Pt: 260
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 1773
ADR: 8 III
Merck Index: 13,04048

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR