(±)-2-Ethyl-1-hexanol zS, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2-Ethylhexan-1-ol

MQ200

8.00990.0100 8.00990.1000 8.00990.2500 8.00990.9025
8.00990.0100EA 23 GBP
8.00990.0100 8.00990.1000 8.00990.2500 8.00990.9025
(±)-2-Ethyl-1-hexanol zS, Sigma-Aldrich®
(±)-2-Ethyl-1-hexanol
Formula: CH₃(CH₂)₃CH(C₂H₅)CH₂OH
MW: 130.23 g/mol
Boiling Pt: 183
Melting Pt: –76 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 81
Storage Temperature: Ambient
MDL Number: MFCD00004746
CAS Number: 104-76-7
EINECS: 203-234-3
REACH: 01-2119487289-20
Merck Index: 13,03842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc81f9275fbd3a7f8fb0e453419eab1652a4fdec&vid=1f7059eb6b27d30ac2c7e6b83f6bc0dff6002fe7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 1.1 g/l (20 °C)
Melting Point -76 °C
Molar Mass 130.23 g/mol
Boiling Point 184 °C (1013 hPa)
Vapor Pressure 0.13 hPa (20 °C)
Flash Point 75 °C DIN 51758
Refractive Index 1.4317 (20 °C, 589 nm)
Explosion Limit 1.1 - 12.7 %(V)
Density 0.832 g/cm3 (20 °C)
pH 7 (1 g/l, H2O, 20 °C)
Ignition Point 270 - 330 °C DIN 51794
Synonyms Isocotyl alcohol, Isooctanol
Hill Formula C8H18O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR