Bovine aprotinin
Supplier: G Biosciences
786-046
786-046EA 1020 GBP
786-046
Bovine aprotinin
Proteins and Peptides
Aprotinin is a broad range, competitive and reversible inhibitor of chymotrypsin, plasmin, trypsin, kallikrein and other serine proteases. Activity: approx 6,000 KIU/mg.