(R)-(+)-α-Methylbenzylamine, Supelco®

Supplier: Merck
Danger

Synonyms: (R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine

MQ100

77879-25ML
77879-25MLEA 302 GBP
77879-25ML
(R)-(+)-α-Methylbenzylamine, Supelco®
(R)-(+)-α-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 180
Melting Pt: 32.5
Density: 0.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064405
CAS Number: 3886-69-9
EINECS: 223-423-4
Merck Index: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27249b36db4a87e4aeacd7011eb5e61117fa3f55&vid=7a106e137a33e80f5b68bc9d1bda3befce50154a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR