Automation Tips for Beckman Coulter® Biomek® i-Series, FX/NX and Multimek Workstations
Supplier: FLUOTICS
BFXI-30.F BFXI-30.F/A BFXI-30.NS BFXI-30.NS/A BFXI-30.ST BFXI-30.ST/A BFXI-50.F BFXI-50.F/A BFXI-50.NS BFXI-50.NS/A BFXI-50.ST BFXI-50.ST/A BFXI-250.F BFXI-250.NS BFXI-250.ST
736-0952EA 3750 GBP
736-0952 736-0953 736-0954 736-0955 736-0956 736-0957 736-0958 736-0959 736-0960 736-0961 736-0962 736-0963 736-1084 736-1085 736-1086
Automation Tips for Beckman Coulter® Biomek® i-Series, FX/NX and Multimek Workstations
Pipette Tips
Fluotics manufactures a selection of automation tips compatible with Beckman Coulter® Biomek® i-Series, FX/NX, and Multimek workstations. These automation tips are directly interchangeable with their Beckman automation tip counterpart, with no change to the labware definitions required.
  • Certified RNase-/DNase-free and nonpyrogenic
  • Produced from virgin, high grade polypropylene
  • Available in non sterile, sterile and barrier filter formats
Compatible with Biomek® i-Series, FX/NX and Multimek workstations with 384 heads. Ultra polishing techniques ensureMore Product Information