Microplate washers, AquaMax®
Supplier: MOLECULAR DEVICES
AQUAMAX2KBUNDL AQUAMAX4KBUNDL
735-0382EA 0 GBP
735-0382 735-0383
Microplate washers, AquaMax®
Microplate Washers
AquaMax® microplate washers are configurable with user interchangeable heads for both 96- and 384-well microplate washing applications, including plate or cell washing.
  • Wash heads can be changed in 30 seconds with no tools using innovative head latch mechanism
  • All replacement heads are recognised by the washer base, no recalibration required
  • Simultaneous well washing without plate indexing or quadrant processing
  • Completely programmable via touch screen interface
  • No external pumps or computer
  • Comprehensive, automated cleaning utilities
  • Robotics-friendly
More Product Information