Microplate readers, SpectraMax® Plus384
Supplier: MOLECULAR DEVICES
PLUS384 PLUS384SCACAD
735-0346EA 0 GBP
735-0346 735-0353
Microplate readers, SpectraMax® Plus384
Microplate Readers
SpectraMax® Plus 384 microplate reader can run both standard spectrophotometer and microplate reader applications on the same instrument. SpectraMax® Plus 384 provides ultrafast, full spectral range detection for cuvettes, 96-well, and 384-well microplates, with the only temperature-independent method for pathlength correction. Temperature control and shaking included as standard.
  • Monitoring of temperature-sensitive reactions with consistent temperature regulation from ambient to 45 °C
  • Thorough mixing of samples in microplate wells with user-programmable
More Product Information