VWR®, 3D Cubic Bags
Supplier: VWR Collection
734-2960EA 3640 GBP
734-2960 734-2961
VWR®, 3D Cubic Bags
Bags Media Bags
These 3D cubic bags can fit in existing support containers, but can also be supplied together with a matching container made of plastic or stainless steel.
3D cubic bags are available in 200 and 500 L volume with 2 ports on top of the bag. Both ports consisting of silicone platinum tubing with MPX-connectors.
VWR® 3D cubic bags are made from a proprietary film, which features a polyethylene inner and outer layer, and a high oxygen barrier layer. The film has high clarity and flexibility, and is resistant to a wide range of chemicals.
Certifications: The bag film is compliant to EP 3.More Product Information