Microcarriers, Corning®
Supplier: Corning
3781 3772 3779 3786 4620 4621 4622 4623 4624
734-2909EA 230 GBP
734-2909 734-2907 734-2908 734-2911 734-2913 734-2914 734-2915 734-2916 734-2917
Microcarriers, Corning®
Beads Microcarriers
Corning® Microcarriers are designed to simplify bioprocessing scale up. The microcarriers are sterile, ready to use, and available in closed systems packaging to be used directly with bioreactors. Additionally, the microcarriers are surface treated to enhance cell attachment, maximising cell yield and viability.
  • USP Class VI polystyrene material provides a consistent platform
  • Available with a choice of surface treatments
  • Sterile (SAL 10⁻⁶)
  • Certified non pyrogenic
  • Available as closed system solutions
Corning® Microcarriers are available with a choiceMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk