Cell strainers, Corning®
Supplier: Corning
431750 431751 431752
734-2760EA 94 GBP
734-2760 734-2761 734-2762
Cell strainers, Corning®
Cell Strainers
Corning® cell strainers are sterile, easy to use devices for rapidly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Fits 50 ml Falcon® conical tubes.
  • Strong nylon mesh for optimal performance in a variety of applications
  • Extended lip on strainer enables aseptic handling with forceps
  • Moulded colour-coded polypropylene frame with tab enables easy handling and identification
  • Sterilised by gamma irradiation, non cytotoxic
  • Conveniently accessible in individual packaging
You can use these versatile devices toMore Product Information