SC substrate, Corning® Synthemax® II
Supplier: Corning
3535
734-2634EA 550 GBP
734-2634
SC substrate, Corning® Synthemax® II
Cell Culture Supplements and Additives
Synthemax® II-SC Substrate is an easy to use, self-coating material for creating a unique synthetic surface that mimics the natural cell environment. This surface supports the adhesion and expansion of stem cells in their undifferentiated state and directs differentiation into specialised cell types. In addition, it offers a synthetic, xeno-free alternative to biological coatings and/or feeder cell layers used in hES and iPS cell culture, whenever needed and appropriate.
Synthemax® II-SC Substrate allows for the scalable multi-expansion of pluripotent stem cells in defined media with:
Stable doubling time
Phenotypic marker expression (Oct4, SSEA4)
Retention of pluripotency
Retention of normal karyotype
Synthemax® II-SC Substrate is compatible with plastic and glass surfaces, lot-traceable, gamma sterilised and quality tested for lot-to-lot consistency.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Species
Source
Environmental certification RECs, ISO 14001 - Kennebunk