Chamber slides, Nunc™ Lab-Tek™
Supplier: Thermo Fisher Scientific
177402 177380 177372 178599 155380 155361 155383 155411 177445 177437 177429 177399 177410
734-2122EA 208 GBP
734-2122 734-2120 734-2119 734-2127 734-2058 734-2056 734-2060 734-2062 734-2126 734-2125 734-2124 734-2121 734-2766
Chamber slides, Nunc™ Lab-Tek™
Slides Chamber Slides
PS medium chamber on glass or Permanox™ plastic slide.
  • Cells grow on a standard microscope slide
  • No cell transfer needed prior to visualisation/staining
  • Upper chamber can be removed when culturing is complete
Suitable for use with fluorescent labels.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Environmental certification IRECS; ISO14001 (Certificate Registration No. EMS 752603) NAR Certificate of Retirement