Corning® Culture Dishes, Nontreated, Sterile, Corning
Supplier: Corning
431111 431272 431301
734-1728EA 210 GBP
734-1728 734-1731 734-1732
Corning® Culture Dishes, Nontreated, Sterile, Corning
Culture Dishes Bioassay Dishes
Optically clear PS. For applications where cell attachment is not desired.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilized by gamma radiation
  • Certified non-pyrogenic
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Oneonta