24-well permeable supports system, polystyrene, sterile, HTS Transwell®
Supplier: Corning
3396 3397 3398 3399 3378 3379
734-1559EA 427 GBP
734-1559 734-1560 734-1561 734-1562 734-4072 734-4073
24-well permeable supports system, polystyrene, sterile, HTS Transwell®
Microplates HTS Microplates
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
  • Cell growth area is 0,33 cm²/well
  • Choice of either polyester (PET) membrane (0,4 µm pore size) or polycarbonate (PC) membrane (0,49 m and 3,0 µm pore sizes)
  • Treated for optimal cell attachment
  • Sterile
The HTS Transwell-24 permeable support has an array of 24 wellsMore Product Information