Angiogenesis system, Corning® BioCoat®
Supplier: Corning
354150 354149 354144 354142
734-1023EA 0 GBP
734-1023 734-1022 734-1021 734-1019
Angiogenesis system, Corning® BioCoat®
Microplates Insert Plates and Inserts
The Corning® BioCoat® Angiogenesis systems facilitate investigation of compound effects on endothelial cell invasion, migration, and tubulogenesis. The availability of these standardised assays has facilitated a better understanding of the molecular mechanism of angiogenesis and simplified the routine use of cell based assays for screening of anti- and pro-angiogenic compounds.
  • Address key steps in the angiogenesis process using standardised cell-based assays
  • Increase throughput with screening-compatible formats and data acquisition options
  • Obtain
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable