Dialysis membrane Spectra/Por® 1
Supplier: Spectrum Laboratories
132645 132650 132655 132660 132665 132670 132675
734-0662EA 475 GBP
734-0662 734-0663 734-0665 734-0666 734-0667 734-0669 734-0670
Dialysis membrane Spectra/Por® 1
Tubing Dialysis Tubing
MWCO 6 to 8 kD, regenerated cellulose.
  • Transparent and flexible
This economical membrane is ideal for a broad range of applications in which there is a significant size difference between the MW species being separated; including sample prep, desalting, buffer exchange, pH change, protein purification. Offering a good chemical compatibility, Standard Grade RC can be used with dilute strong acids and bases, concentrated weak acids and bases, most alcohols and some mild or dilute organics, including DMSO. Standard RC can tolerate pH 2 to 12 andMore Product Information