Collagen I-coated plates, Corning® BioCoat®
Supplier: Corning
354400 356400 354408 354500 354505 356408 356500 356505
734-0108EA 97.4 GBP
734-0108 734-0274 734-0115 734-0166 734-0170 734-0277 734-0295 734-0296
Collagen I-coated plates, Corning® BioCoat®
Microplates Cell Culture Plates
PS coated with rat tail collagen type I. Collagen is an integral part of the framework that holds cells and tissues together and has been recognised as a useful matrix for improving cell culture. In vitro use of collagen can exert effects on the adhesion, morphology, growth, migration, and differentiation of a variety of cell types. Applications include promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, studies of the effects of collagen I on cell behaviour, improving survival of primary cell lines in culture, and cellMore Product Information