VWR®, Red Taq DNA polymerase

Supplier: VWR Chemicals

733-1323EA 429 GBP
733-1323 733-1834 733-2409 733-2408
VWR®, Red Taq DNA polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits

Red Taq DNA Polymerase is a blend of Taq DNA polymerase combined with an inert red dye. The dye enables quick visual recognition of reactions to which enzyme has been added, as well as confirmation of complete mixing.


  • Ideal choice for routine applications
  • High performance, thermostable DNA polymerase
  • Optimal for TA cloning


Taq DNA polymerase concentration: 5 Units/μl


10X Key Buffer: Tris-HCl pH 8,5; (NH₄)₂SO₂, 15 mM MgCl₂, 1% Tween® 20


EU = Units


Delivery information: VWR® Taq DNA Polymerase is usually supplied with Key Buffer. Key Buffer (NH⁴⁺) gives a superior amplification signal (high yield) minimising the need for optimisation of the Mg²⁺ concentration, or the annealing temperature in most primer-template systems.

Order Now

SPECIFICATIONS

Description Taq RED DNA Polymerase 5 U/µl 2.500 units Taq RED DNA Polymerase 5 U/µl 10.000 units Taq RED DNA Polymerase 5 U/µl 1.000 units Taq RED DNA Polymerase 5 U/µl 500 units
Environmentally Preferable
Format Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer
Features Visible dye Visible dye Visible dye Visible dye
Hot start No No No No
Concentration 5 U/µl 5 U/µl 5 U/µl 5 U/µl
Units 2500 10000 1000 500
Size 5×0,1 ml + 15×1,5 ml 3×0,667 ml + 18×5 ml 2×0,1 ml + 6×1,5 ml 1×0,1 ml + 3×1,5 ml
Buffer composition 10X Key Buffer, 10X Extra Buffer and 25 mM MgCl₂ 10X Key Buffer, 10X Extra Buffer and 25 mM MgCl₂ 10X Key Buffer, 10X Extra Buffer and 25 mM MgCl₂ 10X Key Buffer, 10X Extra Buffer and 25 mM MgCl₂
High fidelity No No No No
Amplicon size ≤5 kb ≤5 kb ≤5 kb ≤5 kb
Processivity Standard Standard Standard Standard
Exonuclease activity 5’→3’ 5’→3’ 5’→3’ 5’→3’
Product overhang 3'A 3'A 3'A 3'A
Storage conditions – 20 °C (long-term), + 4 °C (6 months) – 20 °C (long-term), + 4 °C (6 months) – 20 °C (long-term), + 4 °C (6 months) – 20 °C (long-term), + 4 °C (6 months)
Application Standard amplification Standard amplification Standard amplification Standard amplification

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR