Deep well storage plates, 96-well, 2,2 ml, Mark II
Supplier: THERMO MATRIX TECHNOLOGIES
AB-0932
732-4910EA 286 GBP
732-4910
Deep well storage plates, 96-well, 2,2 ml, Mark II
Microplates Deep Well Microplates
PP. 96-well plates with round wells for storage and screening applications.
  • 2,2 ml working volume when sealed with adhesive or heat seals or 1,8 ml when sealed with 96-cap sealing mat
  • Conical well bottom for ease of sample retrieval
  • Raised rim design with support and stacking ring
Colour: Natural.
Colour: Natural