VWR®, Real-Time PCR Plates, 96-Well and 384-Well
Supplier: VWR Collection
211-0297EA 294 GBP
211-0297 211-0300 211-0301 211-0302 211-0303 211-0304 211-0305 211-0315 211-0317 732-3422 732-3486 732-3489 732-3490 732-3491 732-3492 732-3495 732-3496
VWR®, Real-Time PCR Plates, 96-Well and 384-Well
Microplates PCR Plates
VWR® 96- and 384-well Real-Time PCR plates are compatible with most leading Real-Time PCR instruments including most BioRad, Applied Biosystems, Roche Lightcycler and Analytik Jena models.
  • Ultra-thin walls to optimise thermal transfer resulting in increased reaction efficiencies
  • Available with ultra-clear wells for easy visualisation of small sample volumes
  • Available in white options which increase the intensity of fluorescence transmitted through the cap or sealing film
  • Made of virgin polypropylene which is naturally low binding without the need to
More Product Information